Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
248434
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
21
2152
NM_001034955.1
NP_001030127.1
21
2218
NM_001034956.1
NP_001030128.1
18
1705
NM_001034957.1
NP_001030129.1
13
1270
NM_001290294.1
NP_001277223.1
21
2201
NM_001290295.1
NP_001277224.1
16
1484
NM_001290296.1
NP_001277225.1
19
2062
NM_001290297.1
NP_001277226.1
16
1483
NM_001290298.1
NP_001277227.1
17
1588
NM_006434.2
NP_006425.2
13
1270
NM_015385.3
NP_056200.1
17
1853
NM_024991.2
NP_079267.1
17
1580
XM_006717589.1
XP_006717652.1
21
2202
XM_006717593.1
XP_006717656.1
15
1435
XM_011539140.1
XP_011537442.1
23
2946
XM_011539150.1
XP_011537452.1
22
2871
XM_011539155.1
XP_011537457.1
17
2521
XM_011539167.1
XP_011537469.1
16
1804
XM_017015500.1
XP_016870989.1
19
2619
XM_017015501.1
XP_016870990.1
22
2871
XM_017015502.1
XP_016870991.1
21
2778
XM_017015503.1
XP_016870992.1
20
2703
XM_017015504.1
XP_016870993.1
21
1860
XM_017015505.1
XP_016870994.1
19
2619
XM_017015506.1
XP_016870995.1
20
1785
XM_017015507.1
XP_016870996.1
21
2778
XM_017015508.1
XP_016870997.1
19
1692
XM_017015509.1
XP_016870998.1
19
1714
XM_017015510.1
XP_016870999.1
19
2619
XM_017015511.1
XP_016871000.1
18
1608
XM_017015512.1
XP_016871001.1
16
1519
XM_017015513.1
XP_016871002.1
16
1510
XM_017015514.1
XP_016871003.1
17
1594
XM_017015515.1
XP_016871004.1
20
2061
XM_017015516.1
XP_016871005.1
19
2034
XM_017015517.1
XP_016871006.1
19
1977
XM_017015518.1
XP_016871007.1
19
1990
XM_017015519.1
XP_016871008.1
18
1963
XM_017015520.1
XP_016871009.1
18
1902
XM_017015521.1
XP_016871010.1
18
1915
XM_017015522.1
XP_016871011.1
17
1875
XM_017015523.1
XP_016871012.1
17
1831
XM_017015524.1
XP_016871013.1
16
1804
XM_017015525.1
XP_016871014.1
20
1789
XM_017015526.1
XP_016871015.1
19
1692
XM_017015527.1
XP_016871016.1
19
1714
XM_017015528.1
XP_016871017.1
18
1665
XM_017015529.1
XP_016871018.1
18
1617
XM_017015530.1
XP_016871019.1
18
1608
XM_017015531.1
XP_016871020.1
18
1621
XM_017015532.1
XP_016871021.1
17
1590
XM_017015533.1
XP_016871022.1
17
1594
XM_017015534.1
XP_016871023.1
19
1692
XM_017015535.1
XP_016871024.1
17
1546
XM_017015536.1
XP_016871025.1
17
1537
XM_017015537.1
XP_016871026.1
16
1506
XM_017015538.1
XP_016871027.1
18
1665
XM_017015539.1
XP_016871028.1
16
1519
XM_017015540.1
XP_016871029.1
16
1510
GenBank mRNA
AB037717.1
-
17
1512
AF136380.1
-
12
1225
AF136381.1
-
18
2017
AF330623.1
-
15
1438
AF330624.1
-
1
136
AF356525.1
-
21
2163
AF356526.1
-
21
2229
AF356527.1
-
18
1716
AJ489942.1
-
16
1543
AK294924.1
-
5
462
AK300407.1
-
16
1437
AK304130.1
-
5
525
AK304753.1
-
17
2496
AL117472.1
-
17
1811
AM260536.1
-
21
2159
BC042612.1
-
15
1877
BC152463.1
-
17
1512
8862599
BX640814.1
-
5
1515
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0