Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016848.5 | NP_058544.3 | 2 | 824 | ||
XM_011518785.2 | XP_011517087.1 | 2 | 4354 | |||
XM_011518786.2 | XP_011517088.1 | 1 | 31 | |||
XM_017014809.1 | XP_016870298.1 | 2 | 4365 | |||
XM_017014810.1 | XP_016870299.1 | 2 | 4365 | |||
GenBank mRNA | BC026314.1 | - | 2 | 825 | 4819086 | |
D84361.1 | - | 2 | 697 | |||
KU176125.1 | - | 2 | 121 |