Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320724.1 | NP_001307653.1 | 6 | 1686 | ||
NM_016940.2 | NP_058636.1 | 5 | 1581 | |||
GenBank mRNA | AF212232.1 | - | 5 | 1542 | ||
AK000056.1 | - | 5 | 1585 | |||
AK223357.1 | - | 5 | 1592 | |||
BC012546.1 | - | 5 | 1549 | 4291536 | ||
Y19009.1 | - | 5 | 1560 |