Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017423.2 | NP_059119.2 | 1, 2 | 210 | ||
XM_005263062.1 | XP_005263119.1 | 1, 2 | 210 | |||
GenBank mRNA | AJ002744.1 | - | 1, 2 | 159 | ||
BC046129.1 | - | 1, 2 | 188 | 4442080 | ||
BC047468.1 | - | 1, 2 | 185 | 5271749 | ||
DB238519.1 | - | 1, 2 | 210 |