Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243764.1 | NP_001230693.1 | 7 | 1134 | ||
NM_001243765.1 | NP_001230694.1 | 9 | 1742 | |||
XM_005245680.1 | XP_005245737.1 | 26 | 3556 | |||
XM_005245681.2 | XP_005245738.1 | 26 | 3515 | |||
XM_006711693.1 | XP_006711756.1 | 25 | 3358 | |||
XM_011510223.2 | XP_011508525.1 | 27 | 3583 | |||
XM_017002995.1 | XP_016858484.1 | 10 | 2828 | |||
XM_017002996.1 | XP_016858485.1 | 9 | 1311 | |||
XM_017002997.1 | XP_016858486.1 | 9 | 1419 | |||
XM_017002998.1 | XP_016858487.1 | 10 | 8406 | |||
XM_017002999.1 | XP_016858488.1 | 9 | 1219 | |||
GenBank mRNA | AB007940.1 | - | 10 | 1718 | ||
AB019490.1 | - | 13 | 1956 | |||
AB019491.1 | - | 11 | 1425 | |||
AB019492.2 | - | 10 | 1314 | |||
AB019493.1 | - | 11 | 1656 | |||
AK298711.1 | - | 7 | 1100 | |||
AK316355.1 | - | 7 | 1134 | |||
AL035303.1 | - | 7 | 1011 | |||
CR627374.1 | - | 8 | 1175 |