Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243764.1 | NP_001230693.1 | 7 | 1080 | ||
NM_001243765.1 | NP_001230694.1 | 9 | 1688 | |||
XM_005245680.1 | XP_005245737.1 | 26 | 3502 | |||
XM_005245681.2 | XP_005245738.1 | 26 | 3461 | |||
XM_006711693.1 | XP_006711756.1 | 25 | 3304 | |||
XM_011510223.2 | XP_011508525.1 | 27 | 3529 | |||
XM_017002995.1 | XP_016858484.1 | 10 | 2774 | |||
XM_017002996.1 | XP_016858485.1 | 9 | 1257 | |||
XM_017002997.1 | XP_016858486.1 | 9 | 1365 | |||
XM_017002998.1 | XP_016858487.1 | 10 | 8352 | |||
XM_017002999.1 | XP_016858488.1 | 9 | 1165 | |||
GenBank mRNA | AB007940.1 | - | 10 | 1664 | ||
AB019490.1 | - | 13 | 1902 | |||
AB019491.1 | - | 11 | 1371 | |||
AB019492.2 | - | 10 | 1260 | |||
AB019493.1 | - | 11 | 1602 | |||
AK298711.1 | - | 7 | 1046 | |||
AK316219.1 | - | 7 | 1048 | |||
AK316355.1 | - | 7 | 1080 | |||
AL035303.1 | - | 7 | 957 | |||
CR627374.1 | - | 8 | 1121 |