Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052995.2 | NP_443721.1 | 1 | 129 | ||
GenBank mRNA | AF388366.1 | - | 1 | 129 | ||
AF388368.1 | - | 1 | 129 |