Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007167.3 | NP_009098.3 | 2 | 269 | ||
GenBank mRNA | AK299293.1 | - | 2 | 228 | ||
AK310703.1 | - | 2 | 210 | |||
AL389985.1 | - | 2 | 194 | |||
AL705390.1 | - | 2 | 222 | |||
AL832911.1 | - | 2 | 215 | |||
AL833263.1 | - | 2 | 242 | |||
BC033903.1 | - | 2 | 251 | |||
BC068534.1 | - | 2 | 269 | |||
BC107880.1 | - | 2 | 250 | |||
BC117463.1 | - | 2 | 137 | |||
BT007117.1 | - | 1 | 41 |