Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPIDR SIRNA Assays ›
Gene Symbol:
SPIDR
Gene Name:
scaffolding protein involved in DNA repair
Gene Aliases:
KIAA0146
Chromosome Location:
Chr.8: 47260878 - 47736001 on Build GRCh38
UniGene ID:
Hs.381058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
248870
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPIDR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080394.3
NP_001073863.1
4
405
NM_001282916.1
NP_001269845.1
3
338
NM_001282919.1
NP_001269848.1
4
368
NR_104581.1
4
405
XM_005251189.3
XP_005251246.1
4
347
XM_005251191.3
XP_005251248.1
4
347
XM_005251193.3
XP_005251250.1
4
347
XM_005251195.4
XP_005251252.1
4
355
XM_005251198.4
XP_005251255.1
4
376
XM_005251199.4
XP_005251256.1
3
291
XM_006716443.3
XP_006716506.1
4
315
XM_006716444.3
XP_006716507.1
4
339
XM_011517497.2
XP_011515799.1
4
347
XM_011517500.2
XP_011515802.1
4
365
XM_017013268.1
XP_016868757.1
4
347
XM_017013269.1
XP_016868758.1
4
347
XM_017013270.1
XP_016868759.1
4
365
XM_017013271.1
XP_016868760.1
4
462
XM_017013273.1
XP_016868762.1
4
376
XM_017013274.1
XP_016868763.1
4
355
XM_017013277.1
XP_016868766.1
4
332
XM_017013278.1
XP_016868767.1
4
321
XM_017013279.1
XP_016868768.1
3
291
XM_017013280.1
XP_016868769.1
4
376
XM_017013281.1
XP_016868770.1
4
341
XM_017013282.1
XP_016868771.1
4
355
XM_017013283.1
XP_016868772.1
3
291
XM_017013284.1
XP_016868773.1
4
355
XM_017013285.1
XP_016868774.1
4
376
XM_017013286.1
XP_016868775.1
3
291
XM_017013287.1
XP_016868776.1
4
341
XM_017013289.1
XP_016868778.1
1
65
XM_017013290.1
XP_016868779.1
3
291
XM_017013291.1
XP_016868780.1
3
291
GenBank mRNA
AK122820.1
-
5
703
AK293809.1
-
4
328
AK293972.1
-
4
328
AK294248.1
-
2
118
AK294275.1
-
3
260
AK297681.1
-
4
327
AK303579.1
-
4
315
AK309842.1
-
4
341
BC031627.1
-
4
330
D63480.1
-
4
333
DR000775.1
-
4
394
HY012281.1
-
4
406
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
positive regulation of protein complex assembly
regulation of establishment of protein localization to chromosome
cellular response to ionizing radiation
cellular response to hydroxyurea
cellular response to camptothecin
positive regulation of double-strand break repair
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0