Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017523.3 | NP_059993.2 | 2 | 379 | ||
NM_199139.2 | NP_954590.1 | 2 | 379 | |||
NR_046396.1 | 2 | 379 | ||||
NR_046397.1 | 2 | 379 | ||||
NR_046398.1 | 2 | 379 | ||||
XM_005256706.4 | XP_005256763.1 | 2 | 951 | |||
XM_005256708.2 | XP_005256765.1 | 2 | 804 | |||
XM_011523945.2 | XP_011522247.1 | 2 | 174 | |||
XM_011523946.2 | XP_011522248.1 | 2 | 174 | |||
XM_011523948.1 | XP_011522250.1 | 2 | 174 | |||
XM_011523949.2 | XP_011522251.1 | 2 | 174 | |||
XM_017024780.1 | XP_016880269.1 | 2 | 174 | |||
GenBank mRNA | AK056908.1 | - | 2 | 769 | ||
AK290276.1 | - | 3 | 312 | |||
AK292710.1 | - | 2 | 159 | |||
AK292848.1 | - | 2 | 159 | |||
BC032776.1 | - | 2 | 162 | |||
BC058017.1 | - | 2 | 146 | |||
BC073156.1 | - | 2 | 178 | 6068243 | ||
BM475564.1 | - | 2 | 150 | |||
BQ639060.1 | - | 2 | 345 | |||
BX649188.1 | - | 3 | 351 | |||
EF028165.1 | - | 2 | 137 | |||
EF028166.1 | - | 2 | 137 | |||
X99699.1 | - | 2 | 137 |