Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023946.3 | NP_076435.1 | 2 | 151 | ||
NM_177457.4 | NP_803252.1 | 2 | 151 | |||
NM_177476.3 | NP_803429.1 | 2 | 170 | |||
NM_177477.3 | NP_803430.1 | 2 | 284 | |||
GenBank mRNA | AF321824.1 | - | 2 | 110 | ||
BC014560.2 | - | 2 | 126 | |||
BG701653.1 | - | 2 | 170 | |||
BI603715.1 | - | 2 | 289 | |||
BM172253.1 | - | 2 | 169 | |||
BQ685632.1 | - | 2 | 99 | |||
DB501423.1 | - | 2 | 284 | |||
HY110957.1 | - | 2 | 168 |