Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291572.1 | NP_001278501.1 | 18 | 4496 | ||
NM_005899.4 | NP_005890.2 | 21 | 4723 | |||
NM_031862.3 | NP_114068.1 | 21 | 4531 | |||
XM_006721903.1 | XP_006721966.1 | 20 | 4295 | |||
XM_011524813.1 | XP_011523115.1 | 21 | 4832 | |||
XM_017024642.1 | XP_016880131.1 | 22 | 5835 | |||
XM_017024643.1 | XP_016880132.1 | 20 | 4622 | |||
XM_017024644.1 | XP_016880133.1 | 20 | 4321 | |||
GenBank mRNA | AY450308.1 | - | 21 | 4446 | ||
D30756.1 | - | 21 | 4522 |