Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032444.2 | NP_115820.2 | 15 | 7040 | ||
XM_011522715.2 | XP_011521017.1 | 15 | 7051 | |||
XM_017023775.1 | XP_016879264.1 | 12 | 6445 | |||
GenBank mRNA | AB058687.1 | - | 4811 | |||
AL442083.1 | - | 2 | 1580 | |||
BC036335.1 | - | 6 | 4432 |