Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142725.1 | NP_001136197.1 | 3 | 180 | ||
NM_080663.2 | NP_542394.2 | 3 | 180 | |||
GenBank mRNA | AK127391.1 | - | 3 | 183 | ||
AK292594.1 | - | 4 | 393 | |||
AK304347.1 | - | 3 | 179 | |||
BC010503.1 | - | 3 | 180 |