Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009960.1 | NP_001009960.1 | 3 | 452 | ||
NM_001083956.1 | NP_001077425.1 | 4 | 730 | |||
NM_001085368.1 | NP_001078837.1 | 4 | 557 | |||
NM_138494.2 | NP_612503.1 | 3 | 625 | |||
XM_017012598.1 | XP_016868087.1 | 6 | 1630 | |||
XM_017012599.1 | XP_016868088.1 | 6 | 788 | |||
XM_017012600.1 | XP_016868089.1 | 6 | 801 | |||
XM_017012601.1 | XP_016868090.1 | 6 | 787 | |||
XM_017012602.1 | XP_016868091.1 | 5 | 1525 | |||
XM_017012603.1 | XP_016868092.1 | 4 | 1650 | |||
XM_017012604.1 | XP_016868093.1 | 4 | 567 | |||
XM_017012605.1 | XP_016868094.1 | 5 | 578 | |||
XM_017012606.1 | XP_016868095.1 | 3 | 483 | |||
XM_017012607.1 | XP_016868096.1 | 3 | 469 | |||
XM_017012608.1 | XP_016868097.1 | 6 | 1663 | |||
XM_017012609.1 | XP_016868098.1 | 5 | 677 | |||
XM_017012610.1 | XP_016868099.1 | 5 | 624 | |||
GenBank mRNA | AK128057.1 | - | 2 | 1689 | ||
AK314593.1 | - | 3 | 468 | |||
AK316024.1 | - | 4 | 548 | |||
AY099353.1 | - | 3 | 451 | |||
BC037407.1 | - | 3 | 415 | 5752559 | ||
BI548533.1 | - | 4 | 477 | |||
CR749270.1 | - | 3 | 233 | |||
DN993232.1 | - | 4 | 493 |