Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017580.2 | NP_060050.2 | 1 | 557 | ||
XM_005269925.4 | XP_005269982.1 | 2 | 412 | |||
XM_005269926.4 | XP_005269983.1 | 2 | 600 | |||
XM_006717907.2 | XP_006717970.1 | 2 | 1282 | |||
XM_017016357.1 | XP_016871846.1 | 2 | 2013 | |||
XM_017016358.1 | XP_016871847.1 | 1 | 125 | |||
GenBank mRNA | AJ252060.1 | - | 1 | 592 | ||
AK302810.1 | - | 2 | 368 | |||
AK307386.1 | - | 1 | 197 |