Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 6 | 1500 | ||
NM_001164644.1 | NP_001158116.1 | 6 | 1470 | |||
NM_001164645.1 | NP_001158117.1 | 6 | 1356 | |||
NM_001164646.1 | NP_001158118.1 | 6 | 1364 | |||
NM_001164647.1 | NP_001158119.1 | 5 | 1440 | |||
NM_005411.4 | NP_005402.3 | 6 | 1511 | |||
XM_005270062.4 | XP_005270119.1 | 5 | 1500 | |||
XM_006717953.2 | XP_006718016.1 | 6 | 1493 | |||
GenBank mRNA | AK298029.1 | - | 5 | 1400 | ||
BC026229.1 | - | 2 | 983 | |||
HQ021433.2 | - | 5 | 1440 | |||
HQ021434.1 | - | 5 | 1451 | |||
HQ021435.1 | - | 5 | 1440 | |||
HQ021436.1 | - | 5 | 1439 | |||
HQ021437.1 | - | 6 | 1522 | |||
HQ021438.1 | - | 6 | 1511 | |||
HQ021439.1 | - | 6 | 1510 | |||
HQ021440.1 | - | 6 | 1511 | |||
HQ021441.1 | - | 6 | 1500 | |||
HQ021442.1 | - | 6 | 1499 | |||
JX502764.1 | - | 6 | 1511 | |||
JX502765.1 | - | 6 | 1500 |
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 | 1535 | ||
NM_001320813.1 | NP_001307742.1 | 6 | 1538 | |||
NM_001320814.1 | NP_001307743.1 | 5 | 1604 | |||
XM_005270128.3 | XP_005270185.1 | 6 | 1516 | |||
XM_005270132.3 | XP_005270189.1 | 5 | 1503 | |||
XM_011540124.1 | XP_011538426.1 | 6 | 1631 | |||
XM_011540125.1 | XP_011538427.1 | 6 | 1574 | |||
XM_017016608.1 | XP_016872097.1 | 5 | 1427 | |||
GenBank mRNA | AK225599.1 | - | 6 | 1481 | ||
BC139727.1 | - | 4 | 1407 | 40024708 | ||
HQ021421.1 | - | 6 | 1484 | |||
HQ021422.2 | - | 6 | 1484 | |||
HQ021423.2 | - | 6 | 1484 | |||
HQ021424.1 | - | 6 | 1484 | |||
HQ021425.2 | - | 6 | 1484 | |||
HQ021426.1 | - | 6 | 1481 | |||
HQ021427.1 | - | 6 | 1481 | |||
HQ021428.2 | - | 6 | 1481 | |||
HQ021429.2 | - | 6 | 1481 | |||
HQ021430.1 | - | 6 | 1481 | |||
HQ021431.2 | - | 6 | 1481 | |||
HQ021432.1 | - | 6 | 1484 |