Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077697.2 | NP_001071165.2 | 4 | 780 | ||
XM_005272742.4 | XP_005272799.1 | 4 | 755 | |||
XM_011544004.2 | XP_011542306.1 | 4 | 748 |