Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270528.1 | NP_001257457.1 | 8 | 2394 | ||
NM_001270529.1 | NP_001257458.1 | 8 | 2391 | |||
NM_001270530.1 | NP_001257459.1 | 8 | 2251 | |||
NM_017610.7 | NP_060080.6 | 8 | 2394 | |||
XM_005254479.3 | XP_005254536.1 | 8 | 2434 | |||
XM_006720575.3 | XP_006720638.1 | 9 | 2605 | |||
XM_006720579.3 | XP_006720642.1 | 8 | 2251 | |||
XM_011521694.2 | XP_011519996.2 | 8 | 2387 | |||
XM_011521696.2 | XP_011519998.2 | 8 | 2081 | |||
XM_011521697.2 | XP_011519999.2 | 8 | 2081 | |||
XM_011521699.2 | XP_011520001.2 | 8 | 2076 | |||
XM_017022336.1 | XP_016877825.1 | 9 | 2524 | |||
XM_017022337.1 | XP_016877826.1 | 8 | 2431 | |||
XM_017022338.1 | XP_016877827.1 | 8 | 2472 | |||
XM_017022339.1 | XP_016877828.1 | 8 | 2431 | |||
XM_017022340.1 | XP_016877829.1 | 8 | 2431 | |||
XM_017022341.1 | XP_016877830.1 | 8 | 2248 | |||
XM_017022342.1 | XP_016877831.1 | 8 | 2248 | |||
XM_017022343.1 | XP_016877832.1 | 8 | 2248 | |||
GenBank mRNA | AK095327.1 | - | 3 | 473 | ||
AK131304.1 | - | 8 | 2252 | |||
AK131488.1 | - | 7 | 1254 | |||
AL832089.1 | - | 5 | 979 | |||
BC020984.1 | - | 3 | 315 | |||
BC060862.1 | - | 8 | 2394 | 30336202 | ||
BX538130.1 | - | 8 | 2395 | |||
BX647259.1 | - | 8 | 2108 |