Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035267.1 | NP_001030344.1 | 3 | 309 | ||
NM_021104.1 | NP_066927.1 | 4 | 194 | |||
GenBank mRNA | AF026844.1 | - | 1 | 155 | ||
BC014383.1 | - | 1 | 156 | |||
BC015984.1 | - | 1 | 155 | |||
BC017820.1 | - | 3 | 264 | |||
BC032611.1 | - | 1 | 146 | |||
BC062462.1 | - | 1 | 137 | |||
BC064907.1 | - | 1 | 137 | |||
BC066328.1 | - | 3 | 221 | |||
BC070252.1 | - | 1 | 152 | |||
BC070253.1 | - | 1 | 151 | |||
BC070254.1 | - | 1 | 152 | |||
BC070255.1 | - | 1 | 151 | |||
BC070483.1 | - | 1 | 118 | |||
BC082759.1 | - | 3 | 249 | |||
BC102007.1 | - | 4 | 193 | |||
BC102008.1 | - | 4 | 193 | |||
BC102009.1 | - | 4 | 193 | |||
BC105279.1 | - | 1 | 155 | |||
BC105286.1 | - | 1 | 156 | |||
BC105296.1 | - | 1 | 156 | |||
BC105794.1 | - | 1 | 155 | |||
BC105795.1 | - | 1 | 155 | |||
BC105799.1 | - | 1 | 156 | |||
BC142666.1 | - | 1 | 135 | |||
BC142726.1 | - | 1 | 135 | |||
BF673574.1 | - | 4 | 193 | |||
Z12962.1 | - | 4 | 194 |
Gene Symbol: RPL41P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S64030.1 | - | 1 | 73 |