Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 16 | 2010 | ||
NR_027410.1 | 24 | 4258 | ||||
XM_011521596.1 | XP_011519898.1 | 23 | 2855 | |||
XM_011521597.1 | XP_011519899.1 | 22 | 2806 | |||
XM_017022210.1 | XP_016877699.1 | 24 | 3159 | |||
XM_017022211.1 | XP_016877700.1 | 24 | 2928 | |||
XM_017022212.1 | XP_016877701.1 | 23 | 3482 | |||
XM_017022213.1 | XP_016877702.1 | 23 | 2884 | |||
XM_017022214.1 | XP_016877703.1 | 24 | 3156 | |||
XM_017022215.1 | XP_016877704.1 | 24 | 3681 | |||
XM_017022216.1 | XP_016877705.1 | 23 | 4279 | |||
XM_017022217.1 | XP_016877706.1 | 22 | 4201 | |||
XM_017022218.1 | XP_016877707.1 | 24 | 4308 | |||
GenBank mRNA | AK097551.1 | - | 16 | 1999 | ||
BC144610.1 | - | 14 | 1744 | 9053141 | ||
BX647673.1 | - | 23 | 4226 |
Gene Symbol: GOLGA2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120609.1 | 12 | 2874 | |||
GenBank mRNA | AK097852.1 | - | 9 | 1557 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 9 | 1745 | ||
GenBank mRNA | CR936602.1 | - | 12 | 2396 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 9 | 1861 | ||
XM_017022481.1 | XP_016877970.1 | 10 | 1705 | |||
XM_017022482.1 | XP_016877971.1 | 10 | 1705 |
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 10 | 1790 | |||
GenBank mRNA | BX647899.1 | - | 1 | 216 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 8 | 1341 | ||
NM_198181.3 | NP_937824.3 | 9 | 1473 | |||
XM_011521601.2 | XP_011519903.1 | 8 | 2024 | |||
XM_017022226.1 | XP_016877715.1 | 7 | 1460 | |||
GenBank mRNA | AK128268.1 | - | 5 | 1020 | ||
BC041854.1 | - | 8 | 1342 | 5270411 | ||
CR936812.1 | - | 6 | 3876 | |||
DR157204.1 | - | 1 | 217 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 16 | 2010 | ||
NR_027409.1 | 24 | 4272 | ||||
XM_006720440.2 | XP_006720503.1 | 16 | 1975 | |||
XM_017022018.1 | XP_016877507.1 | 20 | 4406 | |||
XM_017022019.1 | XP_016877508.1 | 19 | 4102 | |||
XM_017022020.1 | XP_016877509.1 | 14 | 2097 | |||
GenBank mRNA | AB020662.1 | - | 15 | 1990 | ||
AF163441.1 | - | 14 | 2434 | |||
AF204231.1 | - | 15 | 2327 | |||
AL117569.1 | - | 2 | 317 | |||
AL137686.1 | - | 15 | 1731 | |||
BC144609.1 | - | 14 | 1747 | 9053140 | ||
BC150329.1 | - | 15 | 2001 | |||
BC152410.1 | - | 15 | 1990 | |||
BX648160.1 | - | 23 | 4226 | |||
BX648575.1 | - | 23 | 3979 |
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 18 | 2397 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 18 | 2409 | |||
GenBank mRNA | BC066774.1 | - | 18 | 2383 |