Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020240.1 | - | 2 | 329 | ||
BC041037.1 | - | 2 | 307 | |||
BC073758.1 | - | 2 | 328 | |||
Y14737.1 | - | 2 | 324 |
Gene Symbol: ACSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ496489.1 | - | 202 |
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640854.1 | - | 2 | 339 |
Gene Symbol: IGHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027379.1 | - | 2 | 335 | ||
AK093721.1 | - | 2 | 338 | |||
AK097320.1 | - | 2 | 339 | |||
AK128565.1 | - | 2 | 339 | |||
BC032249.1 | - | 2 | 321 | |||
BC073771.1 | - | 2 | 315 | |||
BX640624.1 | - | 2 | 332 |
Gene Symbol: IGHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC021276.2 | - | 2 | 307 |
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097367.1 | - | 2 | 339 | ||
AK128313.1 | - | 2 | 338 | |||
AK129508.1 | - | 2 | 339 | |||
BC006402.1 | - | 2 | 333 | |||
BC018747.1 | - | 2 | 321 | |||
BC072419.1 | - | 2 | 286 | |||
BX640622.1 | - | 2 | 338 | |||
BX640627.1 | - | 3 | 339 |
Gene Symbol: IGHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062335.1 | - | 2 | 322 |
Gene Symbol: IGHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK301403.1 | - | 2 | 339 |
Gene Symbol: IGHV3-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035024.1 | - | 1 | 202 |
Gene Symbol: IGHV3-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035023.1 | - | 2 | 202 |
Gene Symbol: IGHV3-69-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF460692.1 | - | 1 | 55 | ||
AF460695.1 | - | 55 | ||||
AF460697.1 | - | 55 | ||||
AF460706.1 | - | 1 | 55 | |||
AF460787.1 | - | 55 | ||||
AF460807.1 | - | 55 | ||||
GU552754.1 | - | 112 | ||||
JX420391.1 | - | 1 | 97 | |||
JX420392.1 | - | 1 | 97 |
Gene Symbol: IGHV3-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KP092035.1 | - | 1 | 137 |
Gene Symbol: IGHV4-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640947.1 | - | 2 | 339 |
Gene Symbol: IGLJ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB001733.1 | - | 202 | |||
AB001736.1 | - | 202 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 202 |