Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 18 | 2244 | |||
GenBank mRNA | BC066774.1 | - | 18 | 2218 | ||
BC099913.1 | - | 17 | 3059 | 4838463 | ||
BC125193.1 | - | 17 | 2024 | 40122746 | ||
BC125194.1 | - | 17 | 2024 | 40122749 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 16 | 1845 | ||
NR_027409.1 | 24 | 4107 | ||||
XM_006720440.2 | XP_006720503.1 | 16 | 1810 | |||
XM_017022018.1 | XP_016877507.1 | 20 | 4241 | |||
XM_017022019.1 | XP_016877508.1 | 19 | 3937 | |||
XM_017022020.1 | XP_016877509.1 | 14 | 1932 | |||
GenBank mRNA | AB020662.1 | - | 15 | 1825 | ||
AF163441.1 | - | 14 | 2269 | |||
AF204231.1 | - | 15 | 2162 | |||
AL117569.1 | - | 2 | 152 | |||
AL137686.1 | - | 15 | 1566 | |||
BC093673.1 | - | 16 | 1791 | 7939518 | ||
BC104800.1 | - | 16 | 1791 | 8143803 | ||
BC144609.1 | - | 14 | 1582 | 9053140 | ||
BC150329.1 | - | 15 | 1836 | |||
BC152410.1 | - | 15 | 1825 | |||
BX648160.1 | - | 23 | 4061 | |||
BX648575.1 | - | 23 | 3814 | |||
CR749361.1 | - | 16 | 2998 |
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 16 | 1845 | ||
NR_027410.1 | 24 | 4093 | ||||
XM_011521596.1 | XP_011519898.1 | 23 | 2690 | |||
XM_011521597.1 | XP_011519899.1 | 22 | 2641 | |||
XM_017022210.1 | XP_016877699.1 | 24 | 2994 | |||
XM_017022211.1 | XP_016877700.1 | 24 | 2763 | |||
XM_017022212.1 | XP_016877701.1 | 23 | 3317 | |||
XM_017022213.1 | XP_016877702.1 | 23 | 2719 | |||
XM_017022214.1 | XP_016877703.1 | 24 | 2991 | |||
XM_017022215.1 | XP_016877704.1 | 24 | 3516 | |||
XM_017022216.1 | XP_016877705.1 | 23 | 4114 | |||
XM_017022217.1 | XP_016877706.1 | 22 | 4036 | |||
XM_017022218.1 | XP_016877707.1 | 24 | 4143 | |||
GenBank mRNA | AK097551.1 | - | 16 | 1834 | ||
BC143254.1 | - | 16 | 1791 | 9051756 | ||
BC144610.1 | - | 14 | 1579 | 9053141 | ||
BX647673.1 | - | 23 | 4061 |
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 18 | 2244 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 18 | 2232 | |||
GenBank mRNA | AK292534.1 | - | 18 | 2115 | ||
BC110417.1 | - | 18 | 2203 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 18 | 2232 | |||
GenBank mRNA | BC063309.1 | - | 19 | 2206 | ||
DQ309037.1 | - | 18 | 2206 |
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 18 | 2232 | |||
GenBank mRNA | AK057241.1 | - | 16 | 1710 | ||
AK302625.1 | - | 16 | 1657 | |||
AL833871.1 | - | 7 | 712 | |||
BC144672.1 | - | 15 | 1837 | |||
BC150302.1 | - | 18 | 2206 | 8860399 | ||
DQ309038.1 | - | 16 | 1680 |
Gene Symbol: GOLGA8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282490.1 | NP_001269419.1 | 19 | 1919 |
Gene Symbol: GOLGA8J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282472.1 | NP_001269401.1 | 19 | 1921 | ||
XM_006720651.3 | XP_006720714.1 | 19 | 1941 |
Gene Symbol: GOLGA8M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282468.1 | NP_001269397.1 | 19 | 1921 | ||
XM_005268284.4 | XP_005268341.1 | 19 | 2720 | |||
XM_005268285.4 | XP_005268342.1 | 18 | 2651 | |||
XM_006720654.3 | XP_006720717.1 | 19 | 1864 | |||
XM_011521951.2 | XP_011520253.1 | 20 | 2289 | |||
XM_011521954.2 | XP_011520256.1 | 19 | 1854 |
Gene Symbol: GOLGA8N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282494.1 | NP_001269423.1 | 19 | 1921 |
Gene Symbol: GOLGA8O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277308.1 | NP_001264237.1 | 19 | 1921 | ||
XM_011521988.2 | XP_011520290.1 | 18 | 2537 |
Gene Symbol: GOLGA8R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282484.1 | NP_001269413.1 | 19 | 1918 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 18 | 2188 |
Gene Symbol: GOLGA8T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033933.1 | 17 | 1770 |
Gene Symbol: NXF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK294826.1 | - | 1052 |