Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164644.1 | NP_001158116.1 | 2 | 61 | ||
NM_001164646.1 | NP_001158118.1 | 2 | 61 | |||
NM_005411.4 | NP_005402.3 | 2 | 61 | |||
GenBank mRNA | AK309518.1 | - | 2 | 56 | ||
BC029913.2 | - | 1 | 17 | |||
BI820937.1 | - | 2 | 44 | |||
DA588383.1 | - | 2 | 61 | |||
HQ021437.1 | - | 2 | 61 | |||
HQ021438.1 | - | 2 | 61 | |||
HQ021439.1 | - | 2 | 61 | |||
JX502764.1 | - | 2 | 61 |
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 2 | 115 | ||
NM_001320813.1 | NP_001307742.1 | 2 | 115 | |||
XM_005270128.3 | XP_005270185.1 | 2 | 56 | |||
XM_011540124.1 | XP_011538426.1 | 2 | 211 | |||
XM_011540125.1 | XP_011538427.1 | 2 | 154 | |||
GenBank mRNA | AK225599.1 | - | 2 | 61 | ||
AK298041.1 | - | 2 | 61 | |||
AK298074.1 | - | 2 | 61 | |||
AK298114.1 | - | 2 | 61 | |||
AK313179.1 | - | 2 | 61 | |||
AK316192.1 | - | 2 | 61 | |||
BC111571.1 | - | 2 | 40 | 40069329 | ||
BC157866.1 | - | 2 | 51 | 9057085 | ||
BC157890.1 | - | 2 | 51 | 8862711 | ||
HQ021421.1 | - | 2 | 61 | |||
HQ021422.2 | - | 2 | 61 | |||
HQ021423.2 | - | 2 | 61 | |||
HQ021424.1 | - | 2 | 61 | |||
HQ021425.2 | - | 2 | 61 | |||
HQ021426.1 | - | 2 | 61 | |||
HQ021427.1 | - | 2 | 61 | |||
HQ021428.2 | - | 2 | 61 | |||
HQ021429.2 | - | 2 | 61 | |||
HQ021430.1 | - | 2 | 61 | |||
HQ021431.2 | - | 2 | 61 | |||
HQ021432.1 | - | 2 | 61 |