Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 6 | 862 | ||
NM_001164644.1 | NP_001158116.1 | 6 | 832 | |||
NM_001164645.1 | NP_001158117.1 | 6 | 718 | |||
NM_001164646.1 | NP_001158118.1 | 6 | 726 | |||
NM_001164647.1 | NP_001158119.1 | 5 | 802 | |||
NM_005411.4 | NP_005402.3 | 6 | 873 | |||
XM_005270062.4 | XP_005270119.1 | 5 | 862 | |||
XM_006717953.2 | XP_006718016.1 | 6 | 855 | |||
GenBank mRNA | AK290703.1 | - | 6 | 864 | ||
AK298002.1 | - | 6 | 718 | |||
AK298029.1 | - | 5 | 751 | |||
AK298034.1 | - | 5 | 609 | |||
AK298092.1 | - | 4 | 604 | |||
AK309518.1 | - | 5 | 874 | |||
AK316196.1 | - | 5 | 526 | |||
BC026229.1 | - | 1 | 653 | |||
BC111570.1 | - | 5 | 781 | 40069324 | ||
BC171875.1 | - | 5 | 792 | 9054529 | ||
HQ021433.2 | - | 5 | 802 | |||
HQ021434.1 | - | 5 | 802 | |||
HQ021435.1 | - | 5 | 802 | |||
HQ021436.1 | - | 5 | 802 | |||
HQ021437.1 | - | 6 | 873 | |||
HQ021438.1 | - | 6 | 873 | |||
HQ021439.1 | - | 6 | 873 | |||
HQ021440.1 | - | 6 | 862 | |||
HQ021441.1 | - | 6 | 862 | |||
HQ021442.1 | - | 6 | 862 | |||
JX502764.1 | - | 6 | 873 | |||
JX502765.1 | - | 6 | 862 |
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 | 886 | ||
NM_001320813.1 | NP_001307742.1 | 6 | 889 | |||
NM_001320814.1 | NP_001307743.1 | 5 | 955 | |||
XM_005270128.3 | XP_005270185.1 | 6 | 867 | |||
XM_005270132.3 | XP_005270189.1 | 5 | 854 | |||
XM_011540124.1 | XP_011538426.1 | 6 | 982 | |||
XM_011540125.1 | XP_011538427.1 | 6 | 925 | |||
XM_017016608.1 | XP_016872097.1 | 5 | 778 | |||
GenBank mRNA | AK225599.1 | - | 6 | 832 | ||
AK298041.1 | - | 6 | 739 | |||
AK298074.1 | - | 5 | 558 | |||
AK298114.1 | - | 6 | 685 | |||
AK316192.1 | - | 6 | 765 | |||
BC111571.1 | - | 6 | 811 | 40069329 | ||
BC139727.1 | - | 4 | 758 | 40024708 | ||
BC157866.1 | - | 6 | 825 | 9057085 | ||
BC157890.1 | - | 6 | 825 | 8862711 | ||
HQ021421.1 | - | 6 | 835 | |||
HQ021422.2 | - | 6 | 835 | |||
HQ021423.2 | - | 6 | 835 | |||
HQ021424.1 | - | 6 | 835 | |||
HQ021425.2 | - | 6 | 835 | |||
HQ021426.1 | - | 6 | 832 | |||
HQ021427.1 | - | 6 | 832 | |||
HQ021428.2 | - | 6 | 832 | |||
HQ021429.2 | - | 6 | 832 | |||
HQ021430.1 | - | 6 | 832 | |||
HQ021431.2 | - | 6 | 832 | |||
HQ021432.1 | - | 6 | 835 | |||
K03475.1 | - | 6 | 823 |