Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145472.2 | NP_001138944.1 | 16 | 2457 | ||
NM_001145473.2 | NP_001138945.1 | 2 | 318 | |||
NM_001173129.1 | NP_001166600.1 | 2 | 318 | |||
NM_001308013.1 | NP_001294942.1 | 14 | 2111 | |||
NM_144612.6 | NP_653213.6 | 33 | 5195 | |||
XM_006722388.3 | XP_006722451.1 | 15 | 2291 | |||
XM_006722389.3 | XP_006722452.1 | 16 | 2467 | |||
XM_006722390.3 | XP_006722453.1 | 16 | 2535 | |||
XM_006722391.3 | XP_006722454.1 | 14 | 2105 | |||
XM_011525804.2 | XP_011524106.1 | 22 | 3615 | |||
XM_011525807.2 | XP_011524109.1 | 13 | 2001 | |||
XM_011525810.2 | XP_011524112.1 | 2 | 538 | |||
XM_011525811.2 | XP_011524113.1 | 2 | 1103 | |||
XM_017025540.1 | XP_016881029.1 | 26 | 4411 | |||
XM_017025541.1 | XP_016881030.1 | 27 | 4498 | |||
XM_017025542.1 | XP_016881031.1 | 27 | 4498 | |||
XM_017025543.1 | XP_016881032.1 | 27 | 4498 | |||
XM_017025544.1 | XP_016881033.1 | 27 | 4498 | |||
XM_017025545.1 | XP_016881034.1 | 27 | 4498 | |||
XM_017025548.1 | XP_016881037.1 | 32 | 4845 | |||
GenBank mRNA | AK057232.1 | - | 2 | 305 | ||
AK302484.1 | - | 14 | 2102 | |||
BC041860.1 | - | 16 | 2439 | 5271046 | ||
BC047720.1 | - | 2 | 318 | |||
BX647229.1 | - | 14 | 2111 |