Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSX2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164417.2 | NP_001157889.1 | 3 | 293 | ||
NM_001278701.1 | NP_001265630.1 | 3 | 293 | |||
NM_001278702.1 | NP_001265631.1 | 3 | 293 | |||
GenBank mRNA | BE391023.1 | - | 3 | 247 | ||
CB158955.1 | - | 3 | 210 |
Gene Symbol: SSX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278697.1 | NP_001265626.1 | 3 | 294 | ||
NM_003147.5 | NP_003138.3 | 3 | 294 | |||
NM_175698.2 | NP_783629.1 | 3 | 294 | |||
GenBank mRNA | BC002818.2 | - | 3 | 228 | 3636268 | |
BC007343.2 | - | 3 | 228 | 3615133 | ||
BC016957.1 | - | 3 | 228 | 3916128 | ||
BC069313.1 | - | 2 | 158 | 7262129 | ||
BC071827.1 | - | 3 | 228 | 4641362 | ||
BC103863.2 | - | 3 | 296 | 40003337 | ||
BG104299.1 | - | 3 | 266 | |||
BI560469.1 | - | 3 | 301 | |||
JX014236.1 | - | 2 | 158 | |||
X86175.1 | - | 3 | 249 | |||
Z49105.1 | - | 3 | 256 |
Gene Symbol: SSX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021014.3 | NP_066294.1 | 3 | 298 | ||
XM_011543885.2 | XP_011542187.1 | 3 | 251 | |||
GenBank mRNA | AK310348.1 | - | 3 | 240 | ||
AK310513.1 | - | 3 | 251 | |||
BC005904.1 | - | 3 | 252 | 4064072 | ||
BC103862.1 | - | 3 | 297 | 40003336 | ||
DB444418.1 | - | 3 | 359 | |||
S82471.1 | - | 2 | 158 | |||
U90840.1 | - | 3 | 211 |
Gene Symbol: SSX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021015.3 | NP_066295.3 | 4 | 334 | ||
NM_175723.1 | NP_783729.1 | 3 | 211 | |||
XM_011543949.2 | XP_011542251.1 | 3 | 246 | |||
XM_017029758.1 | XP_016885247.1 | 3 | 246 | |||
GenBank mRNA | BC016640.2 | - | 3 | 300 | 3923927 | |
BE896093.1 | - | 3 | 314 | |||
U90842.1 | - | 2 | 158 |
Gene Symbol: SSX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173358.2 | NP_775494.1 | 3 | 317 | ||
XM_017029430.1 | XP_016884919.1 | 2 | 158 | |||
XM_017029431.1 | XP_016884920.1 | 2 | 158 |
Gene Symbol: SSX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073393.1 | 3 | 245 | |||
GenBank mRNA | DB029485.1 | - | 3 | 245 |