Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 5 | 810 | ||
XM_011525496.1 | XP_011523798.1 | 6 | 612 | |||
XM_017025372.1 | XP_016880861.1 | 5 | 772 | |||
XM_017025373.1 | XP_016880862.1 | 6 | 612 | |||
GenBank mRNA | AJ132583.1 | - | 5 | 540 | ||
AK095877.1 | - | 6 | 576 | |||
AK096709.1 | - | 7 | 597 | |||
AK297107.1 | - | 4 | 314 | |||
AK303037.1 | - | 6 | 605 | |||
AK311414.1 | - | 401 | ||||
BC065294.1 | - | 5 | 499 | 6059589 |
Gene Symbol: LOC101929950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006722258.3 | XP_006722321.1 | 4 | 430 | ||
XM_017025500.1 | XP_016880989.1 | 4 | 435 |
Gene Symbol: LOC440434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036750.2 | 5 | 628 |