Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100112.1 | NP_001093582.1 | 3 | 308 | ||
NM_017534.5 | NP_060004.3 | 3 | 308 | |||
GenBank mRNA | AF111784.1 | - | 1 | 180 | ||
AK300702.1 | - | 3 | 266 | |||
AK308102.1 | - | 3 | 266 | |||
BC093082.1 | - | 2 | 253 | |||
BC126409.1 | - | 2 | 251 | 8992125 | ||
BX510904.2 | - | 3 | 271 | |||
DA765261.1 | - | 3 | 308 | |||
DC379233.1 | - | 3 | 308 |