Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027411.2 | 12 | 1564 |
Gene Symbol: GOLGA2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120609.1 | 6 | 1655 | |||
GenBank mRNA | AK125739.1 | - | 6 | 1655 |
Gene Symbol: GOLGA8DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027407.1 | 12 | 1564 | |||
GenBank mRNA | AK292534.1 | - | 12 | 1447 | ||
BC110417.1 | - | 12 | 1535 | 5171794 |
Gene Symbol: GOLGA8EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033350.1 | 12 | 1564 | |||
GenBank mRNA | BC063309.1 | - | 12 | 1538 | ||
DQ309037.1 | - | 12 | 1538 |
Gene Symbol: GOLGA8F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033351.1 | 12 | 1564 | |||
GenBank mRNA | AK302625.1 | - | 13 | 1265 | ||
AL833871.1 | - | 1 | 44 | |||
BC144672.1 | - | 10 | 1261 | |||
BC150302.1 | - | 12 | 1538 | 8860399 |
Gene Symbol: GOLGA8G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033353.1 | 12 | 1564 | |||
GenBank mRNA | BC066774.1 | - | 12 | 1538 | ||
BC099913.1 | - | 11 | 2391 | 4838463 | ||
BC125193.1 | - | 11 | 1356 | 40122746 | ||
BC125194.1 | - | 11 | 1356 | 40122749 |
Gene Symbol: GOLGA8S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038843.1 | 12 | 1517 |