Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 4 | 474 | ||
XM_005256769.1 | XP_005256826.1 | 4 | 474 | |||
XM_005256770.1 | XP_005256827.1 | 4 | 474 | |||
XM_011523993.1 | XP_011522295.1 | 4 | 474 | |||
XM_011523994.1 | XP_011522296.1 | 4 | 474 | |||
XM_011523995.1 | XP_011522297.1 | 4 | 474 | |||
XM_011523996.1 | XP_011522298.1 | 3 | 378 | |||
XM_011523997.1 | XP_011522299.1 | 2 | 112 | |||
GenBank mRNA | AK315818.1 | - | 4 | 461 | ||
BC073889.1 | - | 4 | 443 | 5222492 | ||
DA426549.1 | - | 4 | 474 |
Gene Symbol: LGALS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002308.3 | NP_002299.2 | 4 | 522 | ||
NM_009587.2 | NP_033665.1 | 4 | 522 | |||
NR_024043.1 | 4 | 451 | ||||
XM_006721892.2 | XP_006721955.1 | 4 | 485 | |||
XM_006721893.2 | XP_006721956.1 | 4 | 514 | |||
XM_006721894.3 | XP_006721957.1 | 4 | 485 | |||
XM_006721895.3 | XP_006721958.1 | 4 | 485 | |||
XM_011524796.1 | XP_011523098.1 | 4 | 514 | |||
XM_017024623.1 | XP_016880112.1 | 4 | 514 | |||
GenBank mRNA | AB003517.1 | - | 4 | 429 | ||
AB005894.1 | - | 4 | 463 | |||
AB006782.1 | - | 4 | 485 | |||
AB008492.1 | - | 4 | 429 | |||
AK126017.1 | - | 4 | 1872 | |||
AK223232.1 | - | 4 | 486 | |||
AK296031.1 | - | 4 | 393 | |||
AK301624.1 | - | 4 | 392 | |||
BC105942.1 | - | 4 | 446 | 40031795 | ||
BC105944.1 | - | 4 | 446 | 40031802 | ||
BC110340.1 | - | 4 | 463 | 5760999 |
Gene Symbol: LGALS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042685.1 | NP_001036150.1 | 4 | 469 | ||
GenBank mRNA | AK290263.1 | - | 4 | 470 | ||
AK296659.1 | - | 3 | 383 | |||
BC105943.1 | - | 4 | 446 | 40031793 | ||
DA668702.1 | - | 4 | 469 |