Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178087.1 | NP_001171558.1 | 1 | 128 | ||
NM_022978.2 | NP_075267.1 | 1 | 128 | |||
GenBank mRNA | DB454907.1 | - | 1 | 128 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 1 | 134 | ||
NM_022968.1 | NP_075257.1 | 1 | 134 | |||
GenBank mRNA | AF073518.1 | - | 1 | 101 | ||
AF073519.1 | - | 1 | 110 | |||
BC021174.1 | - | 1 | 133 | 4824358 | ||
BC035932.1 | - | 1 | 63 | 4701505 | ||
BX107837.1 | - | 1 | 59 | |||
DB444407.1 | - | 1 | 135 |