Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPANXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013453.3 | NP_038481.2 | 1 | 19 | ||
GenBank mRNA | AK225988.1 | - | 1 | 17 | ||
BC054023.1 | - | 1 | 20 | 6648369 | ||
CB115288.1 | - | 1 | 19 |
Gene Symbol: SPANXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145662.3 | NP_663695.1 | 1 | 19 | ||
GenBank mRNA | CB110020.1 | - | 1 | 19 | ||
CB113891.1 | - | 1 | 19 |
Gene Symbol: SPANXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022661.3 | NP_073152.2 | 1 | 52 | ||
GenBank mRNA | AA412605.1 | - | 1 | 15 |
Gene Symbol: SPANXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032417.3 | NP_115793.1 | 1 | 293 | ||
GenBank mRNA | BC005382.1 | - | 1 | 52 | 3935644 | |
BC062672.1 | - | 1 | 233 | 4047937 | ||
BM554791.1 | - | 1 | 293 |