Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029.3 | NP_001020.2 | 3 | 500 | ||
GenBank mRNA | AV681946.1 | - | 3 | 500 | ||
BC002604.1 | - | 3 | 464 | 3143099 | ||
BC013215.2 | - | 1 | 210 | |||
BC015832.1 | - | 1 | 261 | 4807004 | ||
BC070220.1 | - | 1 | 261 | 6388128 | ||
BC105276.1 | - | 1 | 264 | 4287636 | ||
BC105798.1 | - | 1 | 265 | 6726218 | ||
X69654.1 | - | 1 | 258 |
Gene Symbol: RPS26P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002309.1 | 1 | 371 | |||
GenBank mRNA | AW972305.1 | - | 1 | 481 |