Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00680 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125727.1 | 1, 2 | 133 | |||
NR_125728.1 | 1, 2 | 133 | ||||
NR_125729.1 | 1, 2 | 133 | ||||
NR_132994.1 | 1, 2 | 133 | ||||
NR_132995.1 | 1, 2 | 133 | ||||
GenBank mRNA | AL541195.3 | - | 1, 2 | 66 | ||
BC011838.2 | - | 1, 2 | 67 | |||
BC017330.2 | - | 1, 2 | 61 | |||
BC030061.1 | - | 1, 2 | 61 | |||
BC065547.1 | - | 1, 2 | 91 | |||
BI756196.1 | - | 1, 2 | 67 | |||
DA613065.1 | - | 1, 2 | 113 |
Gene Symbol: GUSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027026.1 | 1, 2 | 112 | |||
NR_027027.1 | 1, 2 | 112 | ||||
NR_027028.2 | 1, 2 | 112 | ||||
GenBank mRNA | AK095656.1 | - | 1, 2 | 36 | ||
AK290693.1 | - | 1, 2 | 79 | |||
BC094818.1 | - | 1, 2 | 51 | |||
BC147000.1 | - | 1, 2 | 66 | |||
DA358891.1 | - | 1, 2 | 112 |
Gene Symbol: LINC00680-GUSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003660.2 | 1, 2 | 133 | |||
NR_132997.1 | 1, 2 | 133 | ||||
GenBank mRNA | BC144584.1 | - | 1, 2 | 69 | ||
BM553931.1 | - | 1, 2 | 133 | |||
DA269273.1 | - | 1, 2 | 78 |