Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270528.1 | NP_001257457.1 | 8 | 2688 | ||
NM_001270529.1 | NP_001257458.1 | 8 | 2685 | |||
NM_001270530.1 | NP_001257459.1 | 8 | 2545 | |||
NM_017610.7 | NP_060080.6 | 8 | 2688 | |||
XM_005254479.3 | XP_005254536.1 | 8 | 2728 | |||
XM_006720575.3 | XP_006720638.1 | 9 | 2899 | |||
XM_006720579.3 | XP_006720642.1 | 8 | 2545 | |||
XM_011521694.2 | XP_011519996.2 | 8 | 2681 | |||
XM_011521696.2 | XP_011519998.2 | 8 | 2375 | |||
XM_011521697.2 | XP_011519999.2 | 8 | 2375 | |||
XM_011521699.2 | XP_011520001.2 | 8 | 2370 | |||
XM_017022336.1 | XP_016877825.1 | 9 | 2818 | |||
XM_017022337.1 | XP_016877826.1 | 8 | 2725 | |||
XM_017022338.1 | XP_016877827.1 | 8 | 2766 | |||
XM_017022339.1 | XP_016877828.1 | 8 | 2725 | |||
XM_017022340.1 | XP_016877829.1 | 8 | 2725 | |||
XM_017022341.1 | XP_016877830.1 | 8 | 2542 | |||
XM_017022342.1 | XP_016877831.1 | 8 | 2542 | |||
XM_017022343.1 | XP_016877832.1 | 8 | 2542 | |||
GenBank mRNA | AK095327.1 | - | 3 | 767 | ||
AK131304.1 | - | 8 | 2546 | |||
AK131488.1 | - | 7 | 1548 | |||
AL832089.1 | - | 5 | 1273 | |||
BC020984.1 | - | 3 | 609 | |||
BC060862.1 | - | 8 | 2688 | 30336202 | ||
BX538130.1 | - | 8 | 2689 | |||
BX647259.1 | - | 8 | 2402 |