Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1HL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276687.1 | NP_001263616.1 | 1 | 60 | ||
GenBank mRNA | AF333388.1 | - | 1 | 60 | ||
BC133030.1 | - | 1 | 54 | |||
BC133032.1 | - | 1 | 54 |
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005946.2 | NP_005937.2 | 1 | 87 | ||
GenBank mRNA | AF348995.1 | - | 1 | 69 | ||
AY028617.1 | - | 1 | 87 | |||
BC029475.1 | - | 1 | 84 | 4723818 |
Gene Symbol: MT1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301272.1 | NP_001288201.1 | 1 | 130 | ||
NM_005949.3 | NP_005940.1 | 1 | 130 | |||
GenBank mRNA | BC029453.1 | - | 1 | 130 | 4041046 | |
BC108722.1 | - | 1 | 342 | |||
BM919862.1 | - | 1 | 95 | |||
BQ939879.1 | - | 1 | 75 | |||
W78077.1 | - | 1 | 61 |
Gene Symbol: MT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001447.2 | 1 | 116 | |||
GenBank mRNA | BC070351.1 | - | 1 | 116 | ||
BG747999.1 | - | 1 | 90 |