Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002762.2 | NP_001002762.2 | 4 | 760 | ||
NM_017626.4 | NP_060096.3 | 4 | 760 | |||
XM_005269931.1 | XP_005269988.1 | 4 | 611 | |||
XM_005269932.3 | XP_005269989.1 | 4 | 611 | |||
GenBank mRNA | AK000034.1 | - | 4 | 536 | ||
AK025942.1 | - | 2 | 209 | |||
AK027259.1 | - | 3 | 1237 | |||
AK225051.1 | - | 4 | 536 | |||
AK302071.1 | - | 4 | 841 | |||
BC011812.2 | - | 2 | 129 | |||
BC064920.1 | - | 4 | 526 | 6092812 |