Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3830 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 2896 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3697 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3384 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 3959 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4223 | |||
NM_003879.5 | NP_003870.4 | 10 | 3802 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4697 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3666 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3745 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4632 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6649 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4505 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 3843 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3468 |
Gene Symbol: FKSG48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336875.1 | - | 1 | 205 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336879.1 | - | 1 | 154 | ||
AF336883.1 | - | 1 | 139 | |||
AF336887.1 | - | 1 | 184 | |||
AF338193.1 | - | 1 | 190 | |||
AF338194.1 | - | 1 | 193 | |||
AF338195.1 | - | 1 | 190 | |||
AF338196.1 | - | 1 | 202 | |||
AF338198.1 | - | 1 | 190 | |||
AF338199.1 | - | 1 | 157 | |||
AF469044.1 | - | 1 | 139 |
Gene Symbol: LOC100506290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336880.1 | - | 1 | 211 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 202 | ||
AF336881.1 | - | 1 | 202 | |||
AF338191.1 | - | 1 | 187 |
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127395.2 | NP_001120867.1 | 4 | 2252 | ||
NM_145280.5 | NP_660323.3 | 4 | 2113 |
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 22 | 2836 | ||
GenBank mRNA | AK126330.1 | - | 9 | 2495 | ||
BC063885.1 | - | 22 | 2822 | 30383658 |
Gene Symbol: ORC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024077.1 | - | 4 | 1462 |