Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135821.1 | NP_001129293.1 | 3 | 412 | ||
NM_001135822.1 | NP_001129294.1 | 2 | 269 | |||
NM_001242824.1 | NP_001229753.1 | 2 | 290 | |||
NM_002004.3 | NP_001995.1 | 3 | 467 | |||
XM_005244962.1 | XP_005245019.1 | 2 | 180 | |||
XM_005244963.1 | XP_005245020.1 | 2 | 235 | |||
GenBank mRNA | AK021828.1 | - | 1 | 1203 | ||
AK022841.1 | - | 1 | 1178 | |||
AK291084.1 | - | 3 | 268 | |||
BC010004.2 | - | 2 | 256 | 4132071 | ||
BP370037.1 | - | 2 | 294 | |||
CN346026.1 | - | 2 | 239 | |||
D14697.1 | - | 3 | 362 | |||
DA610405.1 | - | 2 | 235 | |||
J05262.1 | - | 1 | 56 | |||
M29863.1 | - | 1 | 30 |
Gene Symbol: FDPSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003262.1 | 1 | 3896 | |||
GenBank mRNA | BC050292.1 | - | 2 | 2620 |