Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006470.3 | NP_006461.3 | 9 | 1855 | ||
GenBank mRNA | AF096870.1 | - | 6 | 1525 | ||
AK002035.1 | - | 6 | 1321 | |||
AK056026.1 | - | 7 | 2136 | |||
AK314557.1 | - | 6 | 1335 | |||
BC001564.1 | - | 6 | 1185 | |||
BC015674.2 | - | 5 | 874 | |||
BC053514.1 | - | 9 | 1855 | 6263729 | ||
BC067096.1 | - | 9 | 1855 | 6088764 | ||
CR457136.1 | - | 6 | 1298 |
Gene Symbol: TRIM16L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037330.1 | NP_001032407.1 | 5 | 927 | ||
XM_005256479.4 | XP_005256536.1 | 8 | 1101 | |||
XM_005256480.4 | XP_005256537.1 | 9 | 1205 | |||
XM_005256481.4 | XP_005256538.1 | 11 | 1454 | |||
XM_005256483.4 | XP_005256540.1 | 10 | 1346 | |||
XM_011523689.2 | XP_011521991.1 | 6 | 806 | |||
XM_011523690.2 | XP_011521992.1 | 12 | 1516 | |||
XM_017024245.1 | XP_016879734.1 | 13 | 1816 | |||
XM_017024246.1 | XP_016879735.1 | 12 | 1678 | |||
XM_017024247.1 | XP_016879736.1 | 13 | 1819 | |||
XM_017024248.1 | XP_016879737.1 | 12 | 1708 | |||
XM_017024249.1 | XP_016879738.1 | 11 | 1570 | |||
XM_017024250.1 | XP_016879739.1 | 11 | 1571 | |||
XM_017024251.1 | XP_016879740.1 | 11 | 1408 | |||
XM_017024252.1 | XP_016879741.1 | 6 | 1062 | |||
GenBank mRNA | AK293428.1 | - | 9 | 1205 | ||
AK298836.1 | - | 5 | 872 | |||
AK301639.1 | - | 3 | 635 | |||
AK301647.1 | - | 5 | 993 | |||
AK316368.1 | - | 6 | 961 | |||
BC127827.1 | - | 10 | 1276 | 40131713 | ||
BC128591.1 | - | 8 | 1076 | 40127675 | ||
BC146902.1 | - | 5 | 830 | 9056830 | ||
BC146907.1 | - | 5 | 830 | 9056835 | ||
DQ232882.1 | - | 5 | 927 |