Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006234.4 | NP_006225.1 | 3 | 277 | ||
XM_005250452.4 | XP_005250509.1 | 3 | 342 | |||
XM_017012353.1 | XP_016867842.1 | 3 | 274 | |||
GenBank mRNA | AK290515.1 | - | 3 | 274 | ||
BC024165.1 | - | 3 | 243 | 4069069 | ||
BC040116.1 | - | 3 | 238 | |||
BC065711.1 | - | 3 | 255 | 4133200 | ||
BC139902.1 | - | 3 | 240 | 40147225 | ||
BC141830.1 | - | 3 | 239 | 40148671 | ||
BM545237.1 | - | 3 | 395 | |||
L37127.1 | - | 3 | 264 | |||
X82385.1 | - | 3 | 264 | |||
X98433.1 | - | 3 | 231 |
Gene Symbol: POLR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032959.5 | NP_116581.3 | 3 | 328 | ||
GenBank mRNA | AB209398.1 | - | 3 | 266 | ||
AJ277739.1 | - | 3 | 200 | |||
BC056864.1 | - | 3 | 218 |
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097615.2 | NP_001091084.2 | 3 | 300 | ||
XM_017012372.1 | XP_016867861.1 | 3 | 339 | |||
XM_017012373.1 | XP_016867862.1 | 3 | 339 | |||
GenBank mRNA | AF468111.1 | - | 3 | 246 | ||
AJ277741.1 | - | 3 | 200 | |||
DA580738.1 | - | 3 | 262 | |||
DB106772.1 | - | 3 | 301 |
Gene Symbol: POLR2J4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003655.3 | 3 | 287 | |||
GenBank mRNA | BC017250.2 | - | 3 | 263 | ||
BC017341.2 | - | 3 | 251 | |||
BC062722.1 | - | 3 | 246 | |||
BC105584.1 | - | 3 | 269 | |||
BX537878.1 | - | 3 | 242 | |||
DA147922.1 | - | 3 | 287 |