Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100630923 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038967.1 | 3 | 738 | |||
GenBank mRNA | AF178686.1 | - | 2 | 100 | ||
AK098276.1 | - | 4 | 548 | |||
BX459931.2 | - | 3 | 701 |
Gene Symbol: LOC100289561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242713.1 | NP_001229642.1 | 3 | 719 | ||
GenBank mRNA | BU073748.1 | - | 3 | 289 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 3 | 683 | |||
GenBank mRNA | BC046153.2 | - | 3 | 670 | ||
BC101598.1 | - | 2 | 139 | 8069104 | ||
BE938866.1 | - | 1 | 42 |
Gene Symbol: POM121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257190.2 | NP_001244119.1 | 2 | 616 | ||
NM_172020.4 | NP_742017.1 | 1 | 65 | |||
XM_006716196.3 | XP_006716259.1 | 2 | 616 | |||
XM_017012853.1 | XP_016868342.1 | 3 | 668 | |||
XM_017012854.1 | XP_016868343.1 | 3 | 308 | |||
GenBank mRNA | AB014518.1 | - | 2 | 586 |
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099415.2 | NP_001092885.2 | 2 | 490 | ||
GenBank mRNA | BC082993.1 | - | 2 | 473 | ||
BC130587.1 | - | 2 | 197 | |||
BC130589.1 | - | 2 | 197 |