Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006932.2 | NP_001006933.2 | 3 | 593 | ||
NM_001318936.1 | NP_001305865.1 | 4 | 644 | |||
NM_001318938.1 | NP_001305867.1 | 2 | 180 | |||
NM_021135.5 | NP_066958.2 | 2 | 446 | |||
XM_006715549.3 | XP_006715612.1 | 3 | 475 | |||
GenBank mRNA | AB209116.1 | - | 4 | 407 | ||
AK095751.1 | - | 3 | 553 | |||
AK295674.1 | - | 2 | 254 | |||
AK310428.1 | - | 2 | 173 | |||
BC002363.2 | - | 4 | 406 | 2961482 | ||
DC402728.1 | - | 2 | 186 | |||
KJ535087.1 | - | 3 | 211 | |||
L07598.1 | - | 2 | 191 | |||
X85106.1 | - | 4 | 362 |