Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006932.2 | NP_001006933.2 | 5 | 767 | ||
NM_001318936.1 | NP_001305865.1 | 6 | 818 | |||
NM_001318937.1 | NP_001305866.1 | 2 | 475 | |||
NM_001318938.1 | NP_001305867.1 | 4 | 354 | |||
NM_021135.5 | NP_066958.2 | 4 | 620 | |||
XM_006715549.3 | XP_006715612.1 | 5 | 649 | |||
GenBank mRNA | AB209116.1 | - | 6 | 581 | ||
AK095751.1 | - | 5 | 727 | |||
AK295674.1 | - | 4 | 428 | |||
AK310428.1 | - | 4 | 347 | |||
BC002363.2 | - | 6 | 580 | 2961482 | ||
DC402728.1 | - | 4 | 360 | |||
KJ535080.1 | - | 2 | 94 | |||
KJ535087.1 | - | 5 | 385 | |||
L07598.1 | - | 4 | 365 | |||
X85106.1 | - | 6 | 536 |