Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036677.1 | 2 | 152 | |||
GenBank mRNA | AF348994.2 | - | 2 | 152 | ||
BC103839.1 | - | 1 | 721 |
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005946.2 | NP_005937.2 | 2 | 146 | ||
GenBank mRNA | AF348995.1 | - | 2 | 128 | ||
AY028617.1 | - | 2 | 146 |
Gene Symbol: MT1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175617.3 | NP_783316.2 | 2 | 252 | ||
XM_005255956.4 | XP_005256013.1 | 2 | 233 | |||
GenBank mRNA | AF495759.1 | - | 2 | 119 | ||
BC009699.2 | - | 2 | 450 | 3892441 | ||
BC062734.1 | - | 2 | 149 | 6560167 | ||
BC131609.1 | - | 2 | 157 | 40115058 | ||
S68949.1 | - | 2 | 73 |
Gene Symbol: MT1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301272.1 | NP_001288201.1 | 2 | 189 | ||
NM_005949.3 | NP_005940.1 | 2 | 189 | |||
GenBank mRNA | BC029453.1 | - | 2 | 189 | 4041046 | |
BC108722.1 | - | 1 | 989 | |||
BM919862.1 | - | 2 | 154 | |||
BQ939879.1 | - | 2 | 134 | |||
W78077.1 | - | 2 | 120 |
Gene Symbol: MT1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301267.1 | NP_001288196.1 | 2 | 148 | ||
NM_005950.2 | NP_005941.1 | 2 | 145 | |||
GenBank mRNA | BC020757.1 | - | 2 | 148 | 4767886 | |
BC048334.1 | - | 1 | 720 | |||
BM817643.1 | - | 2 | 148 | |||
CD175975.1 | - | 2 | 164 | |||
S68954.1 | - | 2 | 73 |
Gene Symbol: MT1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176870.2 | NP_789846.1 | 2 | 183 | ||
GenBank mRNA | AF136177.1 | - | 2 | 160 | ||
AF479613.1 | - | 2 | 73 | |||
BC028280.1 | - | 2 | 145 | 5191481 | ||
BC103841.1 | - | 2 | 128 | 40002178 | ||
BM918307.1 | - | 2 | 201 |
Gene Symbol: MT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC103840.1 | - | 1 | 127 | ||
BC133653.1 | - | 1 | 127 |
Gene Symbol: MT1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005952.3 | NP_005943.1 | 2 | 144 | ||
GenBank mRNA | AK311284.1 | - | 1 | 740 | ||
BC032131.2 | - | 2 | 133 | 4053295 | ||
BC032338.1 | - | 2 | 131 | 5190391 | ||
CD101947.1 | - | 2 | 148 | |||
S68956.1 | - | 2 | 73 | |||
X76717.1 | - | 2 | 125 |
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005953.3 | NP_005944.1 | 2 | 163 | ||
GenBank mRNA | BC007034.1 | - | 1 | 148 | 4051220 | |
BC070289.1 | - | 1 | 142 | 4051053 | ||
BM746626.1 | - | 2 | 163 | |||
BT007315.1 | - | 2 | 73 | |||
M26637.1 | - | 2 | 80 | |||
S52379.1 | - | 2 | 73 | |||
X97260.1 | - | 2 | 73 |