Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536890.2 | XP_011535192.1 | 2 | 182 | ||
XM_011536892.2 | XP_011535194.1 | 2 | 181 | |||
XM_011536893.2 | XP_011535195.1 | 2 | 182 | |||
XM_011536894.2 | XP_011535196.1 | 2 | 182 | |||
XM_011536895.2 | XP_011535197.1 | 2 | 206 | |||
XM_011536896.2 | XP_011535198.1 | 2 | 182 | |||
XM_017021398.1 | XP_016876887.1 | 2 | 184 | |||
XM_017021399.1 | XP_016876888.1 | 2 | 185 | |||
XM_017021412.1 | XP_016876901.1 | 2 | 182 |
Gene Symbol: F2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM994328.1 | - | 1 | 349 | ||
N35000.1 | - | 1 | 343 |
Gene Symbol: PPIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000057.1 | XP_016855546.1 | 10 | 3381 | ||
XM_017000059.1 | XP_016855548.1 | 10 | 3499 |
Gene Symbol: SPG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF090912.1 | - | 1 | 915 |
Gene Symbol: ZFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724513.3 | XP_006724576.1 | 5 | 675 | ||
XM_006724515.3 | XP_006724578.1 | 5 | 677 | |||
XM_011545578.1 | XP_011543880.1 | 5 | 557 | |||
XM_017029788.1 | XP_016885277.1 | 5 | 598 | |||
XM_017029789.1 | XP_016885278.1 | 4 | 1160 | |||
XM_017029790.1 | XP_016885279.1 | 4 | 565 | |||
XM_017029791.1 | XP_016885280.1 | 3 | 455 | |||
XM_017029792.1 | XP_016885281.1 | 3 | 334 | |||
XM_017029793.1 | XP_016885282.1 | 2 | 132 | |||
XM_017029796.1 | XP_016885285.1 | 3 | 167 | |||
GenBank mRNA | BC136308.1 | - | 4 | 268 | 9020295 | |
BC136309.1 | - | 4 | 268 | 9020296 |