Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160354.1 | NP_001153826.1 | 3 | 1336 | ||
NM_001160355.1 | NP_001153827.1 | 3 | 1057 | |||
NM_017527.3 | NP_059997.3 | 3 | 1089 | |||
GenBank mRNA | AK092545.1 | - | 2 | 2105 | ||
BC001291.1 | - | 3 | 715 |