Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145143.1 | NP_001138615.1 | 4 | 300 | ||
NM_001163646.1 | NP_001157118.1 | 4 | 483 | |||
NM_182537.2 | NP_872343.2 | 3 | 303 | |||
GenBank mRNA | AJ437318.1 | - | 4 | 483 | ||
AY159812.2 | - | 3 | 304 | |||
BC101091.2 | - | 2 | 228 | 40009597 |