Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301272.1 | NP_001288201.1 | 3 | 294 | ||
NM_005949.3 | NP_005940.1 | 3 | 252 | |||
GenBank mRNA | BC029453.1 | - | 3 | 252 | 4041046 | |
BC108722.1 | - | 1 | 1384 | |||
BM919862.1 | - | 3 | 217 | |||
BQ939879.1 | - | 3 | 197 | |||
W78077.1 | - | 3 | 224 |
Gene Symbol: MT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005947.2 | NP_005938.1 | 3 | 191 | ||
GenBank mRNA | AF349000.2 | - | 3 | 190 | ||
AY168638.1 | - | 3 | 192 | |||
BC069421.1 | - | 3 | 186 | 7262703 | ||
BC137478.1 | - | 3 | 158 | 9021478 | ||
BC137479.1 | - | 3 | 158 | 9021479 |
Gene Symbol: MT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC103840.1 | - | 1 | 190 | ||
BC133653.1 | - | 1 | 190 |